Assay Details
| Gene Symbol: EPB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013993102.2 | XP_013848556.1 | 738 | 58 | ||
| XM_021097033.1 | XP_020952692.1 | 738 | 58 | |||
| XM_021097044.1 | XP_020952703.1 | 599 | 58 | |||
More Information
Set Membership: |
|

EPB42
Genomic Map