Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124636.6 | XP_003124684.4 | 632 | 108 | ||
| XM_013995690.2 | XP_013851144.1 | 811 | 108 | |||
More Information
Set Membership: |
|

NMRAL1
Genomic Map