Assay Details
| Gene Symbol: GPAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129347.5 | XP_003129395.1 | 999 | 55 | ||
| XM_005656567.3 | XP_005656624.1 | 915 | 55 | |||
| XM_021101799.1 | XP_020957458.1 | 845 | 55 | |||
| XM_021101800.1 | XP_020957459.1 | 1063 | 55 | |||
| XM_021101801.1 | XP_020957460.1 | 756 | 55 | |||
More Information
Set Membership: |
|

GPAT3
Genomic Map