Assay Details
| Gene Symbol: ZBTB49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128851.6 | XP_003128899.1 | 1601 | 71 | ||
| XM_013989281.2 | XP_013844735.1 | 1625 | 71 | |||
| XM_013989283.2 | XP_013844737.1 | 1464 | 71 | |||
More Information
Set Membership: |
|

ZBTB49
Genomic Map