Assay Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064458.1 | XP_020920117.1 | 121 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021064458.1 transcripts.
Set Membership: |
|

ZBTB18
Genomic Map