Assay Details
| Gene Symbol: ZNF391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099817.1 | XP_020955476.1 | 547 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021099817.1 transcripts.
Set Membership: |
|

ZNF391
Genomic Map