Assay Details
| Gene Symbol: ZNF391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013977501.2 | XP_013832955.1 | 666 | 100 | ||
| XM_021099817.1 | XP_020955476.1 | 669 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977501.2, XM_021099817.1 transcripts.
Set Membership: |
|

ZNF391
Genomic Map