Assay Details
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062922.1 | XP_020918581.1 | 336 | 60 | ||
| XM_021062923.1 | XP_020918582.1 | 336 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TTC36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062922.1 | XP_020918581.1 | 336 | 60 | ||
| XM_021062923.1 | XP_020918582.1 | 336 | 60 | |||
Set Membership: |
|