Assay Details
| Gene Symbol: C1H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092546.1 | XP_020948205.1 | 263 | 85 | ||
| XM_021092553.1 | XP_020948212.1 | 265 | 85 | |||
| XM_021092561.1 | XP_020948220.1 | 297 | 85 | |||
| GenBank mRNA | AK232997.1 | - | 1 - 2 | 252 | 85 | |
| AK350327.1 | - | 1 - 2 | 259 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021092546.1, XM_021092553.1, XM_021092561.1 transcripts.
Set Membership: |
|

C1H18orf25
Genomic Map