Assay Details
| Gene Symbol: ACADSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926297.4 | XP_001926332.2 | 3292 | 108 | ||
| GenBank mRNA | AK233088.1 | - | 1 | 118 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ACADSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926297.4 | XP_001926332.2 | 3292 | 108 | ||
| GenBank mRNA | AK233088.1 | - | 1 | 118 | 108 | |
Set Membership: |
|