Assay Details
| Gene Symbol: OXNAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925300.5 | XP_001925335.4 | 470 | 63 | ||
| XM_021071348.1 | XP_020927007.1 | 809 | 63 | |||
| XM_021071349.1 | XP_020927008.1 | 889 | 63 | |||
| XM_021071350.1 | XP_020927009.1 | 520 | 63 | |||
| XM_021071351.1 | XP_020927010.1 | 842 | 63 | |||
| XM_021071352.1 | XP_020927011.1 | 684 | 63 | |||
| XM_021071353.1 | XP_020927012.1 | 746 | 63 | |||
| XM_021071354.1 | XP_020927013.1 | 309 | 63 | |||
| XM_021071355.1 | XP_020927014.1 | 578 | 63 | |||
| GenBank mRNA | AK233190.1 | - | 4 - 5 | 328 | 63 | |
More Information
Set Membership: |
|

OXNAD1
Genomic Map