Assay Details
| Gene Symbol: PMPCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130241.4 | XP_003130289.1 | 915 | 86 | ||
| GenBank mRNA | AK233201.1 | - | 7 - 8 | 873 | 86 | |
| AK239765.1 | - | 7 - 8 | 884 | 86 | ||
| AK392923.1 | - | 7 - 8 | 887 | 86 | ||
| AY610061.1 | - | 7 - 8 | 881 | 86 | ||
More Information
Set Membership: |
|

PMPCB
Genomic Map