Assay Details
| Gene Symbol: MEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122585.5 | XP_003122633.2 | 1459 | 55 | ||
| XM_005660726.3 | XP_005660783.1 | 1454 | 55 | |||
| XM_005660727.3 | XP_005660784.1 | 1501 | 55 | |||
| GenBank mRNA | AK233230.1 | - | 4 - 5 | 486 | 55 | |
More Information
Set Membership: |
|

MEN1
Genomic Map