Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134122.2 | XP_003134170.1 | 226 | 76 | ||
| XM_005672602.2 | XP_005672659.1 | 298 | 76 | |||
| XM_021076709.1 | XP_020932368.1 | 298 | 76 | |||
| XM_021076710.1 | XP_020932369.1 | 246 | 76 | |||
| XM_021076711.1 | XP_020932370.1 | 312 | 76 | |||
| GenBank mRNA | AK233246.1 | - | 2 - 3 | 232 | 76 | |
| AK397469.1 | - | 3 - 4 | 246 | 76 | ||
More Information
Set Membership: |
|

FAXDC2
Genomic Map