Assay Details
| Gene Symbol: LOC100518109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994302.2 | XP_013849756.2 | 503 | 85 | ||
| XM_021082998.1 | XP_020938657.1 | 729 | 85 | |||
| XM_021082999.1 | XP_020938658.1 | 709 | 85 | |||
| XM_021083000.1 | XP_020938659.1 | 527 | 85 | |||
| GenBank mRNA | AK233274.1 | - | 5 - 6 | 656 | 85 | |
| AK393255.1 | - | 5 - 6 | 737 | 85 | ||
| AK393374.1 | - | 5 - 6 | 656 | 85 | ||
More Information
Set Membership: |
|

LOC100518109
Genomic Map