Assay Details
| Gene Symbol: GCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134883.2 | XP_003134931.2 | 858 | 69 | ||
| XM_013985832.2 | XP_013841286.1 | 820 | 69 | |||
| GenBank mRNA | AK233298.1 | - | 2 - 3 | 373 | 69 | |
More Information
Set Membership: |
|

GCK
Genomic Map