Assay Details
| Gene Symbol: UBQLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064658.1 | XP_020920317.1 | 1152 | 63 | ||
| XM_021064659.1 | XP_020920318.1 | 847 | 63 | |||
| GenBank mRNA | AK233335.1 | - | 3 - 4 | 668 | 63 | |
| AK390226.1 | - | 3 - 4 | 668 | 63 | ||
More Information
Set Membership: |
|

UBQLN1
Genomic Map