Assay Details
Gene Symbol: CCDC115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483603.3 | XP_003483651.1 | 977 | 102 | ||
GenBank mRNA | AK233449.1 | - | 4 - 5 | 966 | 102 | |
AK239401.1 | - | 4 - 5 | 444 | 102 | ||
AK393730.1 | - | 4 - 5 | 982 | 102 | ||
AY610283.1 | - | 4 - 5 | 430 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
