Assay Details
| Gene Symbol: LAPTM4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662724.3 | XP_005662781.1 | 1914 | 68 | ||
| XM_021087809.1 | XP_020943468.1 | 822 | 68 | |||
| GenBank mRNA | AK233467.1 | - | 6 - 7 | 800 | 68 | |
| AK236742.1 | - | 7 - 8 | 754 | 68 | ||
| AK397000.1 | - | 6 - 7 | 799 | 68 | ||
More Information
Set Membership: |
|

LAPTM4A
Genomic Map