Assay Details
| Gene Symbol: TCFL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078229.1 | XP_020933888.1 | 1078 | 65 | ||
| XM_021078231.1 | XP_020933890.1 | 1078 | 65 | |||
| XM_021078233.1 | XP_020933892.1 | 1078 | 65 | |||
| GenBank mRNA | AK233502.1 | - | 2 - 3 | 437 | 65 | |
More Information
Set Membership: |
|

TCFL5
Genomic Map