Assay Details
| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132248.5 | XP_003132296.1 | 1882 | 64 | ||
| XM_005669627.2 | XP_005669684.1 | 1899 | 64 | |||
| XM_005669628.3 | XP_005669685.1 | 1732 | 64 | |||
| XM_005669629.3 | XP_005669686.1 | 1821 | 64 | |||
| XM_013981761.2 | XP_013837215.1 | 1733 | 64 | |||
| XM_021068884.1 | XP_020924543.1 | 2658 | 64 | |||
| GenBank mRNA | AK233520.1 | - | 1 | 814 | 64 | |
More Information
Set Membership: |
|

GLYCTK
Genomic Map