Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131498.6 | XP_003131546.1 | 500 | 70 | ||
| XM_005653921.3 | XP_005653978.1 | 545 | 70 | |||
| XM_013979685.2 | XP_013835139.1 | 446 | 70 | |||
| XM_013979688.2 | XP_013835142.1 | 474 | 70 | |||
| XM_021067031.1 | XP_020922690.1 | 374 | 70 | |||
| XM_021067032.1 | XP_020922691.1 | 373 | 70 | |||
| XM_021067033.1 | XP_020922692.1 | 372 | 70 | |||
| GenBank mRNA | AK233536.1 | - | 3 - 4 | 424 | 70 | |
| AK397547.1 | - | 3 - 4 | 468 | 70 | ||
More Information
Set Membership: |
|

ORMDL3
Genomic Map