Assay Details
| Gene Symbol: SAP30BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131203.5 | XP_003131251.2 | 304 | 69 | ||
| XM_003357968.4 | XP_003358016.1 | 384 | 69 | |||
| GenBank mRNA | AK233585.1 | - | 4 - 5 | 352 | 69 | |
| AK235453.1 | - | 4 - 5 | 351 | 69 | ||
| AK396206.1 | - | 4 - 5 | 374 | 69 | ||
| AK400869.1 | - | 3 - 4 | 302 | 69 | ||
More Information
Set Membership: |
|

SAP30BP
Genomic Map