Assay Details
| Gene Symbol: LOC100520216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980819.2 | XP_013836273.2 | 3111 | 83 | ||
| XM_021068115.1 | XP_020923774.1 | 3104 | 83 | |||
More Information
Set Membership: |
|

LOC100520216
Genomic Map