Assay Details
| Gene Symbol: QSOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081047.1 | XP_020936706.1 | 1564 | 74 | ||
| XM_021081048.1 | XP_020936707.1 | 1534 | 74 | |||
| XM_021081049.1 | XP_020936708.1 | 1868 | 74 | |||
More Information
Set Membership: |
|

QSOX2
Genomic Map