Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129244.5 | XP_003129292.1 | 590 | 84 | ||
| XM_003482454.4 | XP_003482502.1 | 596 | 84 | |||
| XM_013978945.2 | XP_013834399.1 | 578 | 84 | |||
More Information
Set Membership: |
|

AP1AR
Genomic Map