Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665330.3 | XP_005665387.1 | 204 | 86 | ||
| XM_005665331.3 | XP_005665388.1 | 306 | 86 | |||
| XM_013999275.2 | XP_013854729.1 | 204 | 86 | |||
| XM_013999276.2 | XP_013854730.1 | 306 | 86 | |||
More Information
Set Membership: |
|

RNF138
Genomic Map