Assay Details
| Gene Symbol: POLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134888.4 | XP_003134936.2 | 2450 | 77 | ||
| XM_013985828.2 | XP_013841282.1 | 737 | 77 | |||
| XM_021078659.1 | XP_020934318.1 | 1939 | 77 | |||
More Information
Set Membership: |
|

POLM
Genomic Map