Assay Details
| Gene Symbol: ZMPSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096012.1 | XP_020951671.1 | 592 | 75 | ||
| XM_021096013.1 | XP_020951672.1 | 420 | 75 | |||
More Information
Set Membership: |
|

ZMPSTE24
Genomic Map