Assay Details
| Gene Symbol: PLD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128759.6 | XP_003128807.2 | 1479 | 76 | ||
| XM_013989271.2 | XP_013844725.1 | 1317 | 76 | |||
| XM_021081504.1 | XP_020937163.1 | 1668 | 76 | |||
| XM_021081505.1 | XP_020937164.1 | 1662 | 76 | |||
| XM_021081506.1 | XP_020937165.1 | 1599 | 76 | |||
| XM_021081507.1 | XP_020937166.1 | 1541 | 76 | |||
| XM_021081508.1 | XP_020937167.1 | 1548 | 76 | |||
| XM_021081509.1 | XP_020937168.1 | 1349 | 76 | |||
More Information
Set Membership: |
|

PLD4
Genomic Map