Assay Details
| Gene Symbol: LOC100737597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003484168.4 | XP_003484216.2 | 856 | 68 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100737597| Gene Symbol: LOC100737597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003484168.4 | XP_003484216.2 | 856 | 68 | ||