Assay Details
| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076594.1 | XP_020932253.1 | 2018 | 86 | ||
| XM_021076595.1 | XP_020932254.1 | 2111 | 86 | |||
More Information
Set Membership: |
|

RANBP3L
Genomic Map