Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126102.4 | XP_003126150.1 | 623 | 63 | ||
| XM_021091594.1 | XP_020947253.1 | 566 | 63 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126102.4 | XP_003126150.1 | 623 | 63 | ||
| XM_021091594.1 | XP_020947253.1 | 566 | 63 | |||
Set Membership: |
|