Assay Details
| Gene Symbol: PROK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069212.1 | XP_020924871.1 | 313 | 97 | ||
| XM_021069214.1 | XP_020924873.1 | 253 | 97 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PROK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069212.1 | XP_020924871.1 | 313 | 97 | ||
| XM_021069214.1 | XP_020924873.1 | 253 | 97 | |||
Set Membership: |
|