Assay Details
| Gene Symbol: SIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660759.3 | XP_005660816.2 | 1005 | 71 | ||
| XM_021082848.1 | XP_020938507.1 | 2962 | 71 | |||
| XM_021082850.1 | XP_020938509.1 | 1005 | 71 | |||
More Information
Set Membership: |
|

SIPA1
Genomic Map