Assay Details
Gene Symbol: PRDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097561.1 | XP_020953220.1 | 1865 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
