Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126827.4 | XP_003126875.1 | 271 | 99 | ||
XM_005655749.3 | XP_005655806.1 | 271 | 99 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126827.4 | XP_003126875.1 | 271 | 99 | ||
XM_005655749.3 | XP_005655806.1 | 271 | 99 |