Assay Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099584.1 | XP_020955243.1 | 2041 | 67 | ||
| XM_021099585.1 | XP_020955244.1 | 1984 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099584.1 | XP_020955243.1 | 2041 | 67 | ||
| XM_021099585.1 | XP_020955244.1 | 1984 | 67 | |||
Set Membership: |
|