Assay Details
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021070332.1 | XP_020925991.1 | 2195 | 73 | ||
XM_021070333.1 | XP_020925992.1 | 2174 | 73 | |||
XM_021070334.1 | XP_020925993.1 | 2096 | 73 | |||
XM_021070335.1 | XP_020925994.1 | 2075 | 73 | |||
XM_021070336.1 | XP_020925995.1 | 2051 | 73 | |||
XM_021070337.1 | XP_020925996.1 | 2018 | 73 | |||
XM_021070338.1 | XP_020925997.1 | 1997 | 73 | |||
XM_021070339.1 | XP_020925998.1 | 1928 | 73 | |||
XM_021070340.1 | XP_020925999.1 | 2027 | 73 | |||
XM_021070341.1 | XP_020926000.1 | 1915 | 73 | |||
XM_021070342.1 | XP_020926001.1 | 2022 | 73 | |||
XM_021070343.1 | XP_020926002.1 | 1661 | 73 | |||
XM_021070344.1 | XP_020926003.1 | 2075 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
