Assay Details
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013987546.2 | XP_013843000.2 | 149 | 73 | ||
XM_021084345.1 | XP_020940004.1 | 165 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013987546.2, XM_021084345.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
