Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126127.5 | XP_003126175.3 | 2095 | 59 | ||
| XM_005663846.3 | XP_005663903.1 | 2156 | 59 | |||
| XM_013997562.2 | XP_013853016.1 | 2759 | 59 | |||
| XM_013997563.2 | XP_013853017.1 | 797 | 59 | |||
More Information
Set Membership: |
|

NCKAP5L
Genomic Map