Assay Details
| Gene Symbol: ZNF438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064198.1 | XP_020919857.1 | 115 | 84 | ||
| XM_021064199.1 | XP_020919858.1 | 115 | 84 | |||
| XM_021064200.1 | XP_020919859.1 | 115 | 84 | |||
| XM_021064201.1 | XP_020919860.1 | 115 | 84 | |||
| XM_021064202.1 | XP_020919861.1 | 115 | 84 | |||
| XM_021064204.1 | XP_020919863.1 | 117 | 84 | |||
| XM_021064205.1 | XP_020919864.1 | 115 | 84 | |||
| XM_021064206.1 | XP_020919865.1 | 114 | 84 | |||
| XM_021064208.1 | XP_020919867.1 | 115 | 84 | |||
| XM_021064209.1 | XP_020919868.1 | 115 | 84 | |||
| XM_021064210.1 | XP_020919869.1 | 115 | 84 | |||
| XM_021064211.1 | XP_020919870.1 | 115 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021064198.1, XM_021064199.1, XM_021064200.1, XM_021064201.1, XM_021064202.1, XM_021064204.1, XM_021064205.1, XM_021064206.1, XM_021064208.1, XM_021064209.1, XM_021064210.1, XM_021064211.1 transcripts.
Set Membership: |
|

ZNF438
Genomic Map