Assay Details
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131532.4 | XP_003131580.2 | 937 | 58 | ||
| XM_005653947.3 | XP_005654004.2 | 798 | 58 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131532.4 | XP_003131580.2 | 937 | 58 | ||
| XM_005653947.3 | XP_005654004.2 | 798 | 58 | |||
Set Membership: |
|