Assay Details
| Gene Symbol: CMTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094002.1 | XP_020949661.1 | 774 | 114 | ||
| XM_021094003.1 | XP_020949662.1 | 622 | 114 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CMTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094002.1 | XP_020949661.1 | 774 | 114 | ||
| XM_021094003.1 | XP_020949662.1 | 622 | 114 | |||
Set Membership: |
|