Assay Details
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358547.4 | XP_003358595.3 | 1650 | 62 | ||
| XM_013982020.2 | XP_013837474.2 | 1556 | 62 | |||
| XM_021069445.1 | XP_020925104.1 | 1671 | 62 | |||
More Information
Set Membership: |
|

PODXL2
Genomic Map