Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079026.1 | XP_020934685.1 | 3820 | 69 | ||
| XM_021079027.1 | XP_020934686.1 | 3808 | 69 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYNC2I1| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079026.1 | XP_020934685.1 | 3820 | 69 | ||
| XM_021079027.1 | XP_020934686.1 | 3808 | 69 | |||