Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079024.1 | XP_020934683.1 | 2317 | 90 | ||
| XM_021079025.1 | XP_020934684.1 | 2304 | 90 | |||
| XM_021079026.1 | XP_020934685.1 | 2316 | 90 | |||
| XM_021079027.1 | XP_020934686.1 | 2304 | 90 | |||

DYNC2I1
Genomic Map