Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354536.4 | XP_003354584.1 | 549 | 85 | ||
| XM_005653013.3 | XP_005653070.1 | 646 | 85 | |||
| XM_005653015.3 | XP_005653072.1 | 718 | 85 | |||
| XM_013992501.2 | XP_013847955.1 | 603 | 85 | |||
| XM_021086422.1 | XP_020942081.1 | 534 | 85 | |||
More Information
Set Membership: |
|

CD2BP2
Genomic Map