Assay Details
| Gene Symbol: CREB3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122829.6 | XP_003122877.1 | 1990 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CREB3L1| Gene Symbol: CREB3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122829.6 | XP_003122877.1 | 1990 | 109 | ||
Set Membership: |
|