Assay Details
| Gene Symbol: CHRNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483098.4 | XP_003483146.2 | 1460 | 76 | ||
| XM_005669210.3 | XP_005669267.2 | 1307 | 76 | |||
More Information
Set Membership: |
|

CHRNB1
Genomic Map